Gene Transcripts
- Gene Symbol
- MAPK10
- Entrez Gene ID
- 5602
- Gene Name
- mitogen-activated protein kinase 10
- Gene Aliases
- JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
- Chromosome Location
- Chr. 4: 86012296 - 86594117 on Build GRCh38
- UniGene ID
- Hs.125503
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022161.1 |
- |
1 |
918 |
|
View Full Product Details