Gene Transcripts

Gene Symbol
LPCAT4
Entrez Gene ID
254531
Gene Name
lysophosphatidylcholine acyltransferase 4
Gene Aliases
AGPAT7, AYTL3, LPAAT-eta, LPEAT2
Chromosome Location
Chr. 15: 34358888 - 34367196 on Build GRCh38
UniGene ID
Hs.352614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK001119.1 - 1 516

View Full Product Details