Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr. 5: 93615130 - 94111699 on Build GRCh38
UniGene ID
Hs.600086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005272111.4 XP_005272168.1 8 5042
XM_017009962.1 XP_016865451.1 7 4908


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK022140.1 - 1 1686 Chr. 5 - 93860669 - 93863825 [-]

View Full Product Details