Gene Transcripts

Gene Symbol
SLCO4A1
Entrez Gene ID
28231
Gene Name
solute carrier organic anion transporter family member 4A1
Gene Aliases
OATP-E, OATP1, OATP4A1, OATPE, OATPRP1, POAT, SLC21A12
Chromosome Location
Chr. 20: 62642445 - 62690700 on Build GRCh38
UniGene ID
Hs.235782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017027827.1 XP_016883316.1 12 2260
GenBank mRNA AK095985.1 - 1 85

View Full Product Details