Gene Transcripts
- Gene Symbol
- ST6GALNAC3
- Entrez Gene ID
- 256435
- Gene Name
- ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3
- Gene Aliases
- PRO7177, SIAT7C, ST6GALNACIII, STY
- Chromosome Location
- Chr. 1: 76074684 - 76632880 on Build GRCh38
- UniGene ID
- Hs.337040
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130749.1 |
- |
1 |
586 |
|
View Full Product Details