Gene Transcripts
- Gene Symbol
- CARD19
- Entrez Gene ID
- 84270
- Gene Name
- caspase recruitment domain family member 19
- Gene Aliases
- BinCARD, C9orf89
- Chromosome Location
- Chr. 9: 93096123 - 93113295 on Build GRCh38
- UniGene ID
- Hs.434213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042201.1 |
- |
1 |
627 |
|
View Full Product Details