Gene Transcripts

Gene Symbol
NAALADL2
Entrez Gene ID
254827
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
Chromosome Location
Chr. 3: 174439669 - 175809510 on Build GRCh38
UniGene ID
Hs.565848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006713560.2 XP_006713623.1 16 7902
XM_011512612.2 XP_011510914.1 14 11156
XM_011512613.1 XP_011510915.1 14 7964
XM_011512615.2 XP_011510917.1 13 11013
XM_011512616.2 XP_011510918.1 13 8694
XM_017006070.1 XP_016861559.1 18 8344
XM_017006071.1 XP_016861560.1 19 8414
XM_017006072.1 XP_016861561.1 20 8548
XM_017006073.1 XP_016861562.1 18 8147
XM_017006074.1 XP_016861563.1 20 10566
XM_017006075.1 XP_016861564.1 17 9291
XM_017006076.1 XP_016861565.1 17 8084
XM_017006077.1 XP_016861566.1 19 9495
XM_017006078.1 XP_016861567.1 17 9312
XM_017006079.1 XP_016861568.1 15 8788
XM_017006080.1 XP_016861569.1 15 9159
XM_017006081.1 XP_016861570.1 13 11009
XM_017006082.1 XP_016861571.1 15 9007
XM_017006083.1 XP_016861572.1 14 7448
GenBank mRNA AK021754.1 - 1 761

View Full Product Details