Gene Transcripts
- Gene Symbol
- Mir99ahg
- Entrez Gene ID
- 77994
- Gene Name
- Mir99a and Mirlet7c-1 host gene (non-protein coding)
- Gene Aliases
- 2810055G20Rik, D730043B02Rik
- Chromosome Location
- Chr. 16: 77329328 - 77558428 on Build GRCm38
- UniGene ID
- Mm.394688
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033222.1 |
- |
|
914 |
|
AK082961.1 |
- |
|
1414 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB649986.1 |
- |
1 |
340 |
|
Chr. 16 - 77594902 - 77773427 [+] |
View Full Product Details