Gene Transcripts
- Gene Symbol
- Adgrl2
- Entrez Gene ID
- 99633
- Gene Name
- adhesion G protein-coupled receptor L2
- Gene Aliases
- AI450192, Gm619, Lec1, Lphh1, Lphn2, mKIAA0786
- Chromosome Location
- Chr. 3: 148815586 - 149445423 on Build GRCm38
- UniGene ID
- Mm.331434
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082954.1 |
- |
2 |
369 |
|
View Full Product Details