Gene Transcripts
- Gene Symbol
- 2010107E04Rik
- Entrez Gene ID
- 70257
- Gene Name
- RIKEN cDNA 2010107E04 gene
- Gene Aliases
- 9430003J03Rik, AU043134, AV124504, Mp68
- Chromosome Location
- Chr. 12: 111961375 - 111966977 on Build GRCm38
- UniGene ID
- Mm.318
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020400.1 |
- |
1 |
86 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA751014.1 |
- |
1 |
458 |
|
Chr. 12 - 111963401 - 111964176 [-] |
View Full Product Details