Gene Transcripts
- Gene Symbol
- Map3k12
- Entrez Gene ID
- 26404
- Gene Name
- mitogen-activated protein kinase kinase kinase 12
- Gene Aliases
- DLK, MUK, Zpk
- Chromosome Location
- Chr. 15: 102497647 - 102518053 on Build GRCm38
- UniGene ID
- Mm.172897
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK161004.1 |
- |
3 |
790 |
|
View Full Product Details