Gene Transcripts
- Gene Symbol
- Rbms1
- Entrez Gene ID
- 56878
- Gene Name
- RNA binding motif, single stranded interacting protein 1
- Gene Aliases
- 2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1
- Chromosome Location
- Chr. 2: 60750193 - 60963450 on Build GRCm38
- UniGene ID
- Mm.259667
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB589872.1 |
- |
1 |
293 |
|
Chr. 2 - 60750301 - 60751191 [-] |
GenBankmRNA |
CB321580.1 |
- |
|
293 |
|
Chr. - - [-] |
GenBankmRNA |
CB588560.1 |
- |
1 |
269 |
|
Chr. 2 - 60750305 - 60751167 [-] |
View Full Product Details