Gene Transcripts
- Gene Symbol
- Prpf40a
- Entrez Gene ID
- 56194
- Gene Name
- PRP40 pre-mRNA processing factor 40 homolog A (yeast)
- Gene Aliases
- 2810012K09Rik, FBP11, Fnbp11, Fnbp3
- Chromosome Location
- Chr. 2: 53134706 - 53191279 on Build GRCm38
- UniGene ID
- Mm.257474
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012731.1 |
- |
1 |
186 |
|
AK044524.1 |
- |
1 |
1245 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD802998.1 |
- |
1 |
516 |
|
Chr. 2 - 53137732 - 53138404 [-] |
View Full Product Details