Gene Transcripts
- Gene Symbol
- LINC01587
- Entrez Gene ID
- 10141
- Gene Name
- long intergenic non-protein coding RNA 1587
- Gene Aliases
- C4orf6, aC1
- Chromosome Location
- Chr. 4: 5524569 - 5527801 on Build GRCh38
- UniGene ID
- Hs.177972
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000623565.3 |
- |
|
826 |
|
Chr. - - [-] |
GENCODE |
ENST00000504943.1 |
- |
2 |
477 |
|
Chr. 4 - 5527152 - 5527801 [+] |
View Full Product Details