Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024340.1 | 2 | 1780 | ||
GenBank mRNA | BC038741.1 | - | 2 | 1781 | |
BC119749.2 | - | 1 | 656 | ||
BC119750.2 | - | 1 | 656 | ||
CR933684.1 | - | 3 | 1506 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444900.1 | - | 2 | 1780 | Chr. 10 - 59958217 - 59960913 [-] |