Gene Transcripts

Gene Symbol
TPTEP1
Entrez Gene ID
387590
Gene Name
transmembrane phosphatase with tensin homology pseudogene 1
Gene Aliases
psiTPTE22
Chromosome Location
Chr. 22: 16601911 - 16648830 on Build GRCh38
UniGene ID
Hs.474116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK094889.1 - 1 1201
AK097082.1 - 1 1768
BC031001.1 - 3 831
EU541207.1 - 3 777
M27826.1 - 2 304


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000592918.5 - 3 812 Chr. 22 - 16601911 - 16615111 [+]
GenBankmRNA AW957800.1 - 1 273 Chr. 22 - 16614076 - 16614691 [+]
GenBankmRNA R78416.1 - 292 Chr. - - [-]
GenBankmRNA AA307645.1 - 1 274 Chr. 22 - 16614074 - 16614501 [+]
GenBankmRNA BX106546.1 - 2 292 Chr. 22 - 16611873 - 16614751 [+]

View Full Product Details