Gene Transcripts

Gene Symbol
Lrch2
Entrez Gene ID
210297
Gene Name
leucine-rich repeats and calponin homology (CH) domain containing 2
Gene Aliases
AI316885, mKIAA1495
Chromosome Location
Chr. X: 147470375 - 147554103 on Build GRCm38
UniGene ID
Mm.30506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081173.1 NP_001074642.1 18 1977
GenBank mRNA AK122516.1 - 6 1741
BC056180.1 - 12 931

View Full Product Details