Gene Transcripts

Gene Symbol
Phf20
Entrez Gene ID
228829
Gene Name
PHD finger protein 20
Gene Aliases
6820402O20Rik
Chromosome Location
Chr. 2: 156196647 - 156309953 on Build GRCm38
UniGene ID
Mm.427078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172674.2 NP_766262.2 8 1259
GenBank mRNA AK033017.1 - 7 1058
AK038573.1 - 8 1166
AK045309.1 - 8 1178
AK203634.1 - 1 60
BC060121.1 - 8 1139 6413021

View Full Product Details