Gene Transcripts

Gene Symbol
Ythdc1
Entrez Gene ID
231386
Gene Name
YTH domain containing 1
Gene Aliases
A730098D12Rik, C80342, mKIAA1966
Chromosome Location
Chr. 5: 86803980 - 86836659 on Build GRCm38
UniGene ID
Mm.196325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177680.3 NP_808348.2 3 405
GenBank mRNA AK135309.1 - 3 661
BC044650.1 - 2 277
CA317046.1 - 2 217
CN528111.1 - 3 327

View Full Product Details