Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172715.3 | NP_766303.1 | 6 | 917 | |
GenBank mRNA | AK030171.1 | - | 7 | 1102 | |
AK138410.1 | - | 5 | 1077 | ||
AK160261.1 | - | 6 | 862 | ||
BC138228.1 | - | 5 | 670 | 8861247 | |
CN719213.1 | - | 2 | 128 |