Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172715.3 | NP_766303.1 | 9, 10 | 1204 | |
GenBank mRNA | AK030171.1 | - | 10, 11 | 1389 | |
AK138410.1 | - | 8, 9 | 1364 | ||
AK160261.1 | - | 9, 10 | 1149 | ||
BC096769.1 | - | 9, 10 | 1137 | 30101305 | |
BC138228.1 | - | 8, 9 | 957 | 8861247 | |
BC145669.1 | - | 8, 9 | 957 | 40130972 | |
CN719213.1 | - | 5, 6 | 415 |