Gene Transcripts

Gene Symbol
Rpl10
Entrez Gene ID
110954
Gene Name
ribosomal protein L10
Gene Aliases
D0HXS648, DXHXS648, DXHXS648E, QM, QM16
Chromosome Location
Chr. X: 74270816 - 74273135 on Build GRCm38
UniGene ID
Mm.100113
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052835.3 NP_443067.1 5 368
GenBank mRNA AK011022.1 - 5 328
AK012561.1 - 4 325
AK014054.1 - 5 330
AK088777.1 - 5 329
AK144831.1 - 5 328
AK146217.1 - 5 329
AK151416.1 - 5 328
AK151655.1 - 5 328
AK151736.1 - 5 327
AK152123.1 - 5 328
AK152854.1 - 5 328
AK153345.1 - 5 328
AK160618.1 - 5 328
AK166610.1 - 5 327
AK167239.1 - 4 364
AK167966.1 - 5 328
AK168250.1 - 5 327
AK168854.1 - 5 328
AK187585.1 - 3 259
AK187587.1 - 3 221
AK210081.1 - 3 217
BC006068.1 - 4 287
BC024901.1 - 4 289 4224016
BC048872.1 - 4 292 5052784
BC082293.1 - 4 289 6486491
BC083327.1 - 4 289 5684015
BC086917.1 - 5 327 6768088
BC092383.1 - 4 313 5710462
BC098204.1 - 4 313 6410755
BC099437.1 - 4 292 30843551
M93980.1 - 5 327
U89405.1 - 2 197
X75312.1 - 4 320

View Full Product Details