Gene Transcripts

Gene Symbol
Pgr15l
Entrez Gene ID
245526
Gene Name
G protein-coupled receptor 15-like
Gene Aliases
Chromosome Location
Chr. X: 97072596 - 97082104 on Build GRCm38
UniGene ID
Mm.336164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033361.3 NP_001028533.1 5 1165
GenBank mRNA AK136917.1 - 5 1096
AK138339.1 - 5 1166
AY255586.1 - 4 886
BC111868.1 - 1 1963

View Full Product Details