Gene Transcripts

Gene Symbol
PRKACG
Entrez Gene ID
5568
Gene Name
protein kinase cAMP-activated catalytic subunit gamma
Gene Aliases
BDPLT19, KAPG, PKACg
Chromosome Location
Chr. 9: 69012529 - 69014123 on Build GRCh38
UniGene ID
Hs.158029
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002732.3 NP_002723.2 1 288
GenBank mRNA BC039888.1 - 1 288

View Full Product Details