Gene Transcripts
- Gene Symbol
- Olfr92
- Entrez Gene ID
- 258448
- Gene Name
- olfactory receptor 92
- Gene Aliases
- MOR256-29, bM573K1.4
- Chromosome Location
- Chr. 17: 37111042 - 37111980 on Build GRCm38
- UniGene ID
- Mm.377508
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146456.2 |
NP_666667.2
|
1 |
861 |
|
View Full Product Details