Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
320640
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
9530098N22Rik
Chromosome Location
Chr. 4: 112072016 - 112168076 on Build GRCm38
UniGene ID
Mm.253775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178786.4 NP_848901.2 3 447
GenBank mRNA AK028550.1 - 3 498
AK028795.1 - 3 448
AK035748.1 - 3 365
AK162813.1 - 4 462
EF494897.1 - 3 246
EF494898.1 - 3 246

View Full Product Details