Gene Transcripts

Gene Symbol
Mrgprg
Entrez Gene ID
381974
Gene Name
MAS-related GPR, member G
Gene Aliases
Gm1098, Mrgg
Chromosome Location
Chr. 7: 143763710 - 143766993 on Build GRCm38
UniGene ID
Mm.297839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203492.2 NP_987077.1 2 1089
GenBank mRNA BC100302.1 - 2 1089

View Full Product Details