Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001260492.1 | NP_001247421.1 | 14 | 2019 | |
NM_001260493.1 | NP_001247422.1 | 14 | 2019 | ||
NM_001260494.1 | NP_001247423.1 | 12 | 1648 | ||
NM_001260495.1 | NP_001247424.1 | 7 | 1072 | ||
NM_001260496.1 | NP_001247425.1 | 6 | 807 | ||
NM_002906.3 | NP_002897.1 | 14 | 2019 | ||
GenBank mRNA | AK295075.1 | - | 11 | 1364 | |
AK312903.1 | - | 14 | 1921 | ||
AK316061.1 | - | 12 | 1550 | ||
BC047109.1 | - | 14 | 1919 | 5284438 | |
BX648536.1 | - | 4 | 735 | ||
DQ916738.1 | - | 14 | 2019 | ||
DQ916739.1 | - | 6 | 807 | ||
DQ916740.1 | - | 7 | 1072 | ||
DQ916741.1 | - | 14 | 2019 | ||
DQ916742.1 | - | 14 | 2019 | ||
L02320.1 | - | 13 | 1739 |