Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002922.3 | NP_002913.3 | 2 | 228 | |
GenBank mRNA | AF493925.1 | - | 2 | 123 | |
AK093544.1 | - | 2 | 180 | ||
AK303138.1 | - | 2 | 182 | ||
AK315607.1 | - | 2 | 182 | ||
AL039059.1 | - | 2 | 232 | ||
BC015510.1 | - | 2 | 160 | 3916789 | |
BG536972.1 | - | 2 | 228 | ||
BT006668.1 | - | 2 | 123 | ||
EU668340.1 | - | 2 | 160 | ||
X73427.1 | - | 2 | 137 |