Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027000.4 | NP_081276.2 | 17 | 1796 | |
GenBank mRNA | AF325354.1 | - | 17 | 1780 | |
AF348208.1 | - | 17 | 1803 | ||
AK032309.1 | - | 5 | 491 | ||
AK163372.1 | - | 16 | 1616 | ||
AK166711.1 | - | 17 | 1803 | ||
BC005514.1 | - | 17 | 1777 | ||
BC025431.1 | - | 17 | 1801 | 5007174 |