Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284351.2 | NP_001271280.1 | 9 | 1587 | |
NM_004174.3 | NP_004165.2 | 9 | 1614 | ||
GenBank mRNA | BC011555.1 | - | 7 | 1149 | |
BC101669.1 | - | 9 | 1510 | 8069175 | |
BC101671.1 | - | 9 | 1510 | 8069177 | |
BC143328.1 | - | 9 | 1483 | 9051833 | |
BP270851.1 | - | 3 | 245 | ||
S83545.1 | - | 4 | 455 | ||
U28043.1 | - | 9 | 1512 |