Gene Transcripts

Gene Symbol
SLC20A2
Entrez Gene ID
6575
Gene Name
solute carrier family 20 member 2
Gene Aliases
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
Chromosome Location
Chr. 8: 42416462 - 42542213 on Build GRCh38
UniGene ID
Hs.653173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257180.1 NP_001244109.1 7 1450
NM_001257181.1 NP_001244110.1 7 1081
NM_006749.4 NP_006740.1 7 1413
XM_005273613.3 XP_005273670.1 7 1188
XM_005273615.3 XP_005273672.1 7 1188
XM_006716390.3 XP_006716453.1 6 1047
XM_006716391.3 XP_006716454.1 8 4668
XM_017013748.1 XP_016869237.1 7 1304
XM_017013749.1 XP_016869238.1 6 4392
XM_017013750.1 XP_016869239.1 8 1323
XM_017013751.1 XP_016869240.1 8 1197
XM_017013752.1 XP_016869241.1 8 1439
GenBank mRNA AK291202.1 - 7 1450
AK295952.1 - 3 291
AK314774.1 - 7 1082
BC028600.2 - 7 1125 4820601
L20852.1 - 6 999

View Full Product Details