Gene Transcripts
- Gene Symbol
- Mcpt9
- Entrez Gene ID
- 54272
- Gene Name
- mast cell protease 9
- Gene Aliases
- Chromosome Location
- Chr. 15: 35018087 - 35020842 on Build Rnor_6.0
- UniGene ID
- Rn.10651
- Species
- Rattus norvegicus
- Gene Symbol
- LOC691603
- Entrez Gene ID
- 691603
- Gene Name
- similar to mast cell protease 9
- Gene Aliases
- Chromosome Location
- Chr. 15: 34691247 - 34694270 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Mcpt10
- Entrez Gene ID
- 54269
- Gene Name
- mast cell protease 10
- Gene Aliases
- Chromosome Location
- Chr. 15: 34622126 - 34625141 on Build Rnor_6.0
- UniGene ID
- Rn.48655
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017146.1 |
NP_058842.1
|
|
206 |
|
GenBank mRNA |
X68657.1 |
- |
|
206 |
|
- Gene Symbol
- Mcpt8
- Entrez Gene ID
- 29269
- Gene Name
- mast cell protease 8
- Gene Aliases
- RMCP8
- Chromosome Location
- Chr. 15: 34644473 - 34647431 on Build Rnor_6.0
- UniGene ID
- Rn.55163
- Species
- Rattus norvegicus
- Gene Symbol
- Mcpt8l2
- Entrez Gene ID
- 408240
- Gene Name
- mast cell protease 8-like 2
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.154796
- Species
- Rattus norvegicus
View Full Product Details