Gene Transcripts
- Gene Symbol
- SNX1
- Entrez Gene ID
- 6642
- Gene Name
- sorting nexin 1
- Gene Aliases
- HsT17379, VPS5
- Chromosome Location
- Chr. 15: 64094071 - 64144234 on Build GRCh38
- UniGene ID
- Hs.188634
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242933.1 |
NP_001229862.1
|
6, 7 |
782 |
|
NM_003099.4 |
NP_003090.2
|
6, 7 |
782 |
|
NM_148955.3 |
NP_683758.1
|
4, 5 |
587 |
|
XM_005254611.2 |
XP_005254668.1
|
6, 7 |
758 |
|
GenBank mRNA |
AB209013.1 |
- |
6, 7 |
589 |
|
AF065483.1 |
- |
6, 7 |
652 |
|
AF065484.1 |
- |
4, 5 |
457 |
|
AK128179.1 |
- |
6, 7 |
674 |
|
AK222561.1 |
- |
6, 7 |
666 |
|
AK222793.1 |
- |
6, 7 |
655 |
|
AK291752.1 |
- |
6, 7 |
666 |
|
AK310618.1 |
- |
3, 4 |
423 |
|
BC000357.2 |
- |
6, 7 |
662 |
2964409 |
BQ935515.1 |
- |
6, 7 |
656 |
|
BT006983.1 |
- |
6, 7 |
652 |
|
U53225.1 |
- |
6, 7 |
697 |
|
View Full Product Details