Gene Transcripts

Gene Symbol
Pfn1
Entrez Gene ID
64303
Gene Name
profilin 1
Gene Aliases
Chromosome Location
Chr. 10: 57273003 - 57275708 on Build Rnor_6.0
UniGene ID
Rn.1152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022511.2 NP_071956.2 3 497
GenBank mRNA BC062405.1 - 3 497
FQ217736.1 - 3 531
FQ221987.1 - 3 531
FQ227505.1 - 3 532
FQ228171.1 - 3 531
FQ232047.1 - 3 530
FQ232161.1 - 3 487
FQ232713.1 - 3 501
FQ232749.1 - 3 496
FQ235292.1 - 3 532
X96967.1 - 3 380

View Full Product Details