Gene Transcripts

Gene Symbol
Sf3b1
Entrez Gene ID
84486
Gene Name
splicing factor 3b, subunit 1
Gene Aliases
Sap155
Chromosome Location
Chr. 9: 61593396 - 61634510 on Build Rnor_6.0
UniGene ID
Rn.198843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053426.1 NP_445878.1 16 2447
XM_006244933.2 XP_006244995.1 16 2444
XM_006244934.2 XP_006244996.1 13 3352

View Full Product Details