Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079515.2 | NP_001072983.1 | 9 | 992 | |
NM_001287801.1 | NP_001274730.1 | 10 | 1097 | ||
NM_001287802.1 | NP_001274731.1 | 8 | 915 | ||
NM_003193.4 | NP_003184.1 | 9 | 944 | ||
GenBank mRNA | AK292287.1 | - | 9 | 849 | |
AK296185.1 | - | 10 | 1027 | ||
AK298050.1 | - | 10 | 1156 | ||
BC008654.1 | - | 9 | 836 | 3872809 | |
BT007086.1 | - | 8 | 768 | ||
U61232.1 | - | 9 | 897 |