Gene Transcripts

Gene Symbol
Rbm11
Entrez Gene ID
288321
Gene Name
RNA binding motif protein 11
Gene Aliases
Chromosome Location
Chr. 11: 14005701 - 14015723 on Build Rnor_6.0
UniGene ID
Rn.154372
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105898.1 NP_001099368.1 5 576
XM_006247994.2 XP_006248056.1 5 720
XM_006247995.2 XP_006248057.1 5 717

View Full Product Details