Gene Transcripts

Gene Symbol
Mdp1
Entrez Gene ID
290230
Gene Name
magnesium-dependent phosphatase 1
Gene Aliases
Mdp-1, RGD1311147
Chromosome Location
Chr. 15: 34336058 - 34340084 on Build Rnor_6.0
UniGene ID
Rn.19870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106039.1 NP_001099509.1 6 557
XM_008770675.1 XP_008768897.1 5 1545

View Full Product Details