Gene Transcripts

Gene Symbol
Obox1
Entrez Gene ID
691860
Gene Name
oocyte specific homeobox 1
Gene Aliases
Chromosome Location
Chr. 1: 77614984 - 77617758 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228359.1 XP_006228421.1 1 210

View Full Product Details