Gene Transcripts

Gene Symbol
March5
Entrez Gene ID
294079
Gene Name
membrane-associated ring finger (C3HC4) 5
Gene Aliases
Rnf153
Chromosome Location
Chr. 1: 255842639 - 255864131 on Build Rnor_6.0
UniGene ID
Rn.218003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106372.2 NP_001099842.1 2 502
XM_008760367.1 XP_008758589.1 2 508
GenBank mRNA BC158801.1 - 2 502

View Full Product Details