Gene Transcripts

Gene Symbol
Mettl14
Entrez Gene ID
295428
Gene Name
methyltransferase like 14
Gene Aliases
RGD1304822
Chromosome Location
Chr. 2: 227566811 - 227583049 on Build Rnor_6.0
UniGene ID
Rn.40840
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106470.1 NP_001099940.1 7 720
XM_006233266.2 XP_006233328.1 7 780
GenBank mRNA BC166789.1 - 7 626

View Full Product Details