Gene Transcripts

Gene Symbol
Svs1
Entrez Gene ID
297081
Gene Name
seminal vesicle secretory protein 1
Gene Aliases
Chromosome Location
Chr. 4: 78639672 - 78644570 on Build Rnor_6.0
UniGene ID
Rn.56148
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199095.1 NP_954526.1 1 272
XM_006236459.1 XP_006236521.1 1 272

View Full Product Details