Gene Transcripts

Gene Symbol
Bcap29
Entrez Gene ID
298943
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
Chromosome Location
Chr. 6: 50883758 - 50923743 on Build Rnor_6.0
UniGene ID
Rn.19451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006980.1 NP_001006981.1 3 234
XM_006239982.2 XP_006240044.1 4 355
GenBank mRNA BC083576.1 - 3 234

View Full Product Details