Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
299737
Gene Name
netrin 4
Gene Aliases
RGD1565947
Chromosome Location
Chr. 7: 34533543 - 34649330 on Build Rnor_6.0
UniGene ID
Rn.140212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106780.1 NP_001100250.1 7, 8 1672

View Full Product Details