Gene Transcripts

Gene Symbol
Rnf26
Entrez Gene ID
300659
Gene Name
ring finger protein 26
Gene Aliases
Chromosome Location
Chr. 8: 48447113 - 48449311 on Build Rnor_6.0
UniGene ID
Rn.15010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113748.1 NP_001107220.1 1 738
GenBank mRNA BC158712.1 - 1 738

View Full Product Details