Gene Transcripts

Gene Symbol
Nnmt
Entrez Gene ID
300691
Gene Name
nicotinamide N-methyltransferase
Gene Aliases
Chromosome Location
Chr. 8: 52918437 - 52938189 on Build Rnor_6.0
UniGene ID
Rn.15755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106819.1 NP_001100289.1 1 256
XM_008766195.1 XP_008764417.1 1 481
GenBank mRNA FQ218983.1 - 1 220

View Full Product Details