Gene Transcripts

Gene Symbol
Nyx
Entrez Gene ID
302516
Gene Name
nyctalopin
Gene Aliases
RGD1561300
Chromosome Location
Chr. X: 10197547 - 10218583 on Build Rnor_6.0
UniGene ID
Rn.153545
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100967.1 NP_001094437.1 3 391
GenBank mRNA DQ393414.1 - 3 391

View Full Product Details