Gene Transcripts
- Gene Symbol
- RAB7A
- Entrez Gene ID
- 7879
- Gene Name
- RAB7A, member RAS oncogene family
- Gene Aliases
- PRO2706, RAB7
- Chromosome Location
- Chr. 3: 128726136 - 128814798 on Build GRCh38
- UniGene ID
- Hs.684374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004637.5 |
NP_004628.4
|
3, 4 |
405 |
|
GenBank mRNA |
AF498942.1 |
- |
2, 3 |
173 |
|
AK225177.1 |
- |
2, 3 |
159 |
|
AK225691.1 |
- |
2, 3 |
152 |
|
AK290721.1 |
- |
3 |
354 |
|
AK298344.1 |
- |
3, 4 |
328 |
|
AK308934.1 |
- |
3, 4 |
348 |
|
AU122058.1 |
- |
3, 4 |
405 |
|
BC008721.2 |
- |
3, 4 |
348 |
2821435 |
BC013728.1 |
- |
2, 3 |
188 |
3882701 |
BX537775.1 |
- |
1, 2 |
121 |
|
View Full Product Details