Gene Transcripts

Gene Symbol
Nsrp1
Entrez Gene ID
303346
Gene Name
nuclear speckle splicing regulatory protein 1
Gene Aliases
Ccdc55, NSrp70, RGD1309863
Chromosome Location
Chr. 10: 63098115 - 63133188 on Build Rnor_6.0
UniGene ID
Rn.145937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037189.1 NP_001032266.1 7 772
XM_008768018.1 XP_008766240.1 6 1920
GenBank mRNA BC099120.1 - 7 772

View Full Product Details